2-Acetamido-6-hydroxypurine ≥97%
Supplier: Matrix Scientific
Synonyms:
N-2-Acetylguanine, N-Acetylguanine
085916-5G
085916-25G
102846-374EA
300
USD
102846-374
102846-376
2-Acetamido-6-hydroxypurine ≥97%
2-Acetamido-6-hydroxypurine
Hazard: Irritant
Formula:
C₇H₇N₅O₂ |
MDL Number:
MFCD00078201 CAS Number: 19962-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...