2-(2-Hydroxyethyl)-1-methylpyrrolidine ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-2-pyrrolidineethanol

085720-10G 085720-1G
102845-816EA 72.31 USD
102845-816 102845-818
2-(2-Hydroxyethyl)-1-methylpyrrolidine ≥97%
2-(2-Hydroxyethyl)-1-methylpyrrolidine

Hazard: Irritant

Formula: C₇H₁₅NO
MW: 129.20 g/mol
MDL Number: MFCD00003174
CAS Number: 67004-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR