Pyrazinamide ≥97%

Supplier: Matrix Scientific

Synonyms: Pyrazinecarboxamide, Pyrazine-2-carboxamide, 2-Pyrazinecarboxamide

085417-10G 085417-50G
102845-002EA 64.62 USD
102845-002 102845-004
Pyrazinamide ≥97%
Pyrazinamide

Hazard: Irritant

Formula: C₅H₅N₃O
MW: 123.11 g/mol
MDL Number: MFCD00006132
CAS Number: 98-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR