5-(2,6-Dichlorophenyl)-1H-tetrazole ≥97%

Supplier: Matrix Scientific

083809-1G 083809-10G
102840-790EA 353.85 USD
102840-790 102840-792
5-(2,6-Dichlorophenyl)-1H-tetrazole ≥97%
5-(2,6-Dichlorophenyl)-1H-tetrazole

Hazard: Irritant

Formula: C₇H₄Cl₂N₄
MDL Number: MFCD00798373
CAS Number: 50907-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR