Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-chloroimidazo[1,2-a]pyridine-2-carboxylate

083645-1G
102840-310EA 750.77 USD
102840-310
Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate ≥97%
Ethyl 5-chloroimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉ClN₂O₂
MW: 224.65 g/mol
MDL Number: MFCD09864797
CAS Number: 67625-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR