4-Propylphenol ≥97%

Supplier: Matrix Scientific

Synonyms: p-Propylphenol, 4-n-Propylphenol

085010-1G 085010-10G
102844-018EA 143.08 USD
102844-018 102844-020
4-Propylphenol ≥97%
4-Propylphenol

Hazard: Irritant

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 232
Melting Pt: 24
MDL Number: MFCD00002395
CAS Number: 645-56-7
UN: 3145
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR