4-Bromobenzamidine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromophenylamidine hydrochloride, 4-Bromobenzimidamide hydrochloride, 4-Bromobenzene-1-carboximidamide hydrochloride

084954-1G 084954-5G
102843-826EA 16.92 USD
102843-826 102843-828
4-Bromobenzamidine hydrochloride ≥95%
4-Bromobenzamidine hydrochloride

Hazard: Irritant

Formula: C₇H₈BrClN₂
MDL Number: MFCD00464967
CAS Number: 55368-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR