2-Amino-m-cresol ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Hydroxy-6-methylaniline, 2-Amino-3-hydroxytoluene, 2-Amino-3-methylphenol, 1-Amino-2-hydroxy-6-methylbenzene
084892-1G
084892-10G
102843-662EA
72.31
USD
102843-662
102843-664
2-Amino-m-cresol ≥97%
2-Amino-m-cresol
Hazard: Irritant
Formula:
C₇H₉NO MW: 123.15 g/mol |
MDL Number:
MFCD00075082 CAS Number: 2835-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...