4-Chlorobenzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

084430-10G 084430-50G
102842-392EA 2087.69 USD
102842-392 102842-394
4-Chlorobenzenesulfonyl chloride ≥95%
4-Chlorobenzenesulphonyl chloride

Hazard: Irritant

Formula: C₆H₄Cl₂O₂S
MDL Number: MFCD00007439
CAS Number: 98-60-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR