α,2,6-Trichlorotoluene ≥97%
Supplier: Matrix Scientific
Synonyms:
2,6-Dichlorobenzyl Chloride
, 1,3-Dichloro-2-(chloromethyl)benzene
, 2,6-Dichlorobenzyl chloride
, 1-(Chloromethyl)-2,6-dichlorobenzene
084341-50G
084341-10G
102842-142EA
64.62
USD
102842-142
102842-144
α,2,6-Trichlorotoluene ≥97%
α,2,6-Trichlorotoluene
Hazard: Irritant
Formula:
C₇H₅Cl₃ MW: 195.47 g/mol |
MDL Number:
MFCD00000897 CAS Number: 2014-83-7 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...