Cyclohexylméthanol ≥97%

Supplier: Matrix Scientific

084280-1G 084280-10G
102841-964EA 56.92 USD
102841-964 102841-966
Cyclohexylméthanol ≥97%
Cyclohexylméthanol

Hazard: Irritant

Formula: C₇H₁₄O
Boiling Pt: 180…181 °C
Density: 0.926
Flash Pt: 71 °C
MDL Number: MFCD00001510
CAS Number: 100-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR