6-Acetoxy-2-naphthoic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Acetoxy-2-naphthoic Acid
082942-1G
082942-10G
102838-486EA
241.54
USD
102838-486
102838-488
6-Acetoxy-2-naphthoic acid ≥97%
6-Acetoxy-2-naphthoic acid
Hazard: Irritant
Formula:
C₁₃H₁₀O₄ MW: 230.22 g/mol |
MDL Number:
MFCD00059556 CAS Number: 17295-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...