1-Methylcyclohexanecarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-1-cyclohexanecarboxylic acid, 1-Methylcyclohexane-1-carboxylic acid

082874-1G 082874-10G 082874-50G
102838-312EA 18.46 USD
102838-312 102838-314 102838-316
1-Methylcyclohexanecarboxylic acid ≥97%
1-Methylcyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₄O₂
MDL Number: MFCD00001463
CAS Number: 1123-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR