N-Acetyl-L-hydroxyproline ≥97%

Supplier: Matrix Scientific

Synonyms: Ac-Hyp-OH

082841-1G 082841-10G
102838-232EA 38.46 USD
102838-232 102838-234
N-Acetyl-L-hydroxyproline ≥97%
N-Acetyl-L-hydroxyproline

Hazard: Irritant

Formula: C₇H₁₁NO₄
MW: 173.168 g/mol
MDL Number: MFCD00037339
CAS Number: 33996-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR