Captopril ≥97%

Supplier: Matrix Scientific

Synonyms: (S)-(-)-1-(3-Mercapto-2-methyl-1-oxopropyl)-L-proline, (S)-(-)-1-(3-Mercapto-2-methylpropionyl)-L-proline, (S)-1-((S)-3-mercapto-2-methylpropanoyl)pyrrolidine-2-carboxylic acid

082839-1G 082839-10G
102838-226EA 15.38 USD
102838-226 102838-228
Captopril ≥97%
Captopril

Purity: 97% CAS: 62571-86-2 MDL Number: MFCD00168073 Molecular Formula: C9H15NO3S Molecular Weight: 217,29 Hazard: Irritant

Formula: C₉H₁₅NO₃S
MW: 217.28 g/mol
MDL Number: MFCD00168073
CAS Number: 62571-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR