Nicotinic acid (Niacin) ≥97%

Supplier: Matrix Scientific

Synonyms: Pyridine-3-carboxylic acid3-Pyridinecarboxylic acidVitamin B33-Picolinic acidNicotinic acidNiacin

082487-50G
102837-444EA 61.54 USD
102837-444
Nicotinic acid (Niacin) ≥97%
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
MW: 123.11 g/mol
MDL Number: MFCD00006391
CAS Number: 59-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR