3-Chloro-4-methylbenzylamine ≥97%

Supplier: Matrix Scientific

081813-1G 081813-10G
102835-880EA 170.77 USD
102835-880 102835-882
3-Chloro-4-methylbenzylamine ≥97%
3-Chloro-4-methylbenzylamine

Hazard: Irritant

Formula: C₈H₁₀ClN
MDL Number: MFCD00014816
CAS Number: 67952-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR