3-Aminophthalimide ≥97%

Supplier: Matrix Scientific

081500-1G 081500-10G
102835-086EA 298.46 USD
102835-086 102835-088
3-Aminophthalimide ≥97%
3-Aminophthalimide

Hazard: Irritant

Formula: C₈H₆N₂O₂
MDL Number: MFCD00041853
CAS Number: 2518-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR