6-Aminophthalide ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Aminoisobenzofuran-1(3H)-one, 6-AMINOPHTHALIDE

081454-1G 081454-10G
102834-974EA 255.38 USD
102834-974 102834-976
6-Aminophthalide ≥97%
6-Aminophthalide

Hazard: Irritant

Formula: C₈H₇NO₂
MW: 149.15 g/mol
MDL Number: MFCD00033530
CAS Number: 57319-65-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR