5-Amino-2-methylindole ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-5-indolamine, 2-Methylindol-5-ylamine, 5-Amino-2-methyl-1H-indole

081379-1G 081379-10G
102834-750EA 61.54 USD
102834-750 102834-752
5-Amino-2-methylindole ≥97%
5-Amino-2-methylindole

Hazard: Irritant

Formula: C₉H₁₀N₂
MW: 146.19 g/mol
MDL Number: MFCD02093426
CAS Number: 7570-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR