2-Aminoquinolin-8-ol

Supplier: Matrix Scientific

Synonyms: 2-Amino-8-quinolinol

080657-500MG 080657-1G 080657-5G
102670-610EA 184.62 USD
102670-610 102670-612 102670-614
2-Aminoquinolin-8-ol
2-Aminoquinolin-8-ol

Hazard: Irritant

Formula: C₉H₈N₂O
MDL Number: MFCD00168931
CAS Number: 70125-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR