1,2-Dihydroacenaphthylene-5-carbaldehyde

Supplier: Matrix Scientific

Synonyms: 1,2-dihydroacenaphthylene-5-carbaldehyde, 5-Acenaphthenecarboxaldehyde

077085-500MG 077085-1G 077085-5G
102664-402EA 258.46 USD
102664-402 102664-404 102664-406
1,2-Dihydroacenaphthylene-5-carbaldehyde
1,2-Dihydroacenaphthylene-5-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀O
MW: 182.22 g/mol
MDL Number: MFCD00805838
CAS Number: 5345-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR