Ethyl-1-acetylcyclobutane-1-carboxylate ≥95%

Supplier: Matrix Scientific

077026-500MG 077026-1G
102664-232EA 356.92 USD
102664-232 102664-234
Ethyl-1-acetylcyclobutane-1-carboxylate ≥95%
Ethyl-1-acetylcyclobutane-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₄O₃
MDL Number: MFCD07787010
CAS Number: 126290-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR