5-Amino-6-chloroquinoline ≥95%

Supplier: Matrix Scientific

076236-1G 076236-5G
102545-134EA 427.69 USD
102545-134 102545-136
5-Amino-6-chloroquinoline ≥95%
5-Amino-6-chloroquinoline

Hazard: Irritant

Formula: C₉H₇ClN₂
MW: 178.6200 g/mol
MDL Number: MFCD02947117
CAS Number: 341010-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR