Dess-Martin periodinane ≥95%

Supplier: Matrix Scientific

Synonyms: 1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one, DMP, (1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one, Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl ester

076222-100G 076222-25G
102545-064EA 223.08 USD
102545-064 102552-024
Dess-Martin periodinane ≥95%
Dess-Martin periodinane

Cannot ship with flammable liquids


Hazard: Oxidizer/Stored under Argon/Store in Freezer/Moisture Sensitive

Formula: C₁₃H₁₃IO₈
Melting Pt: 130…132 °C
MDL Number: MFCD00130127
CAS Number: 87413-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR