Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulfonate ≥95%
Supplier: Matrix Scientific
Synonyms:
(-)-3-Bromocamphor-8-sulphonic acid Ammonium salt, (-)-3-Bromocamphor-8-sulfonic acid Ammonium salt
076215-1G
076215-5G
076215-10G
102545-026EA
49.23
USD
102545-026
102545-028
102545-030
Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulfonate ≥95%
Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulfonate
Hazard: Irritant
Formula:
C₁₀H₁₈BrNO₄S |
MDL Number:
MFCD00151070 CAS Number: 55870-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...