2-Methoxyphenyl-2-(2-(1-methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl)acetamido)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: N-[2-[1-Methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl]acetyl]glycine 2-Methoxyphenyl Ester, Amtolmetin guacil

076179-250MG 076179-1G
102544-856EA 924.62 USD
102544-856 102544-858
2-Methoxyphenyl-2-(2-(1-methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl)acetamido)acetate ≥95%
2-Methoxyphenyl-2-(2-(1-methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl)acetamido)acetate

Hazard: Irritant

Formula: C₂₄H₂₄N₂O₅
MW: 420.47 g/mol
MDL Number: MFCD00866153
CAS Number: 87344-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR