(+)-Cinchonine ≥95%

Supplier: Matrix Scientific

Synonyms: Cinchonine(S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol

076110-10G 076110-100G 076110-5G
102544-540EA 55.38 USD
102544-540 102544-544 102544-542
(+)-Cinchonine ≥95%
(+)-Cinchonine

Hazard: Irritant

Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
MDL Number: MFCD00064372
CAS Number: 118-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR