Tetraethyl orthosilicate (TEOS) ≥95%

Supplier: Matrix Scientific

Synonyms: Tetraethoxysilane

076014-10G 076014-100G
102544-102EA 36.92 USD
102544-102 102544-104
Tetraethyl orthosilicate (TEOS) ≥95%
Tetraethyl orthosilicate (TEOS)

Hazard: Irritant

Formula: Si(OC₂H₅)₄
Density: 0.93
MDL Number: MFCD00009062
CAS Number: 78-10-4
UN: 1292
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR