N-(1-Naphthyl)ethylenediamine dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine
075980-5G
075980-10G
075980-100G
102543-930EA
55.38
USD
102543-930
102543-932
102543-934
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥95%
N-2-Aminoethyl-1-naphthylamine dihydrochloride
Purity: 95+%
CAS: 1465-25-4
MDL Number: MFCD00012556
Molecular Formula: C12H16Cl2N2
Molecular Weight: 259,18
Hazard: Irritant
Formula:
C₁₂H₁₆Cl₂N₂ |
MDL Number:
MFCD00012556 CAS Number: 1465-25-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...