Sulfanilic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

075891-10G 075891-100G
102543-492EA 36.92 USD
102543-492 102543-494
Sulfanilic acid ≥95%
Sulphanilic acid

Hazard: Irritant

Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363
Melting Pt: 279
MDL Number: MFCD00007886
CAS Number: 121-57-3
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR