3-Aminobenzyl alcohol ≥95%
Supplier: Matrix Scientific
075834-100G
075834-25G
102543-234EA
184.62
USD
102543-234
102833-878
3-Aminobenzyl alcohol ≥95%
3-Aminobenzyl alcohol
Hazard: Irritant
Formula:
C₇H₉NO Melting Pt: 91…95 °C |
MDL Number:
MFCD00007817 CAS Number: 1877-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...