4-Amino-3,5-difluorobenzonitrile ≥95%

Supplier: Matrix Scientific

075825-1G 075825-5G
102543-190EA 509.23 USD
102543-190 102543-192
4-Amino-3,5-difluorobenzonitrile ≥95%
4-Amino-3,5-difluorobenzonitrile

Hazard: Irritant

Formula: C₇H₄F₂N₂
MW: 154.1169 g/mol
MDL Number: MFCD03407467
CAS Number: 110301-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR