1,3-Diamino-2-propanol ≥95%

Supplier: Matrix Scientific

Synonyms: 1,3-Diaminopropan-2-ol1,3-Diamino-2-hydroxypropane

075706-100G
102542-632EA 138.46 USD
102542-632
1,3-Diamino-2-propanol ≥95%
1,3-Diamino-2-propanol

Hazard: Irritant

Formula: C₃H₁₀N₂O
MW: 90.13 g/mol
Boiling Pt: 235 °C
Melting Pt: 40…45 °C
Flash Pt: >110 °C
MDL Number: MFCD00008142
CAS Number: 616-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR