D-(-)-Arabinose ≥95%

Supplier: Matrix Scientific

Synonyms: D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal

075554-10G 075554-100G
102541-882EA 364.62 USD
102541-882 102541-884
D-(-)-Arabinose ≥95%
D(-)-Arabinose

Hazard: Irritant

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Melting Pt: 160
MDL Number: MFCD00135608
CAS Number: 10323-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR