L(+)-Arabinose ≥95%

Supplier: Matrix Scientific

Synonyms: Pectin SugarPectinose

075542-100G
102541-832EA 123.08 USD
102541-832
L(+)-Arabinose ≥95%
L(+)-Arabinose

Hazard: Irritant

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
MDL Number: MFCD00067709
CAS Number: 5328-37-0
https://us.vwr.com/stibo/web/std.lang.all/24/52/2302452.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR