3-Chlorophenyl acetylene ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Chloro-3-ethynylbenzene, 3-Chlorophenylacetylene, (3-Chlorophenyl)acetylene
075416-1G
075416-5G
075416-10G
102541-254EA
15.38
USD
102541-254
102541-256
102541-258
3-Chlorophenyl acetylene ≥95%
3-Chlorophenyl acetylene
Hazard: Irritant
Formula:
C₈H₅Cl Boiling Pt: 178…179 °C Density: 1.109 Flash Pt: 62.78 °C |
MDL Number:
MFCD03839984 CAS Number: 766-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...