Thymidine (2'-deoxythymidine) ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, dT, Thymine deoxyriboside, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione, Pyrimidine deoxynucleoside, Thymidine kinase, Thymidine-2-deoxythymidine

075406-5G 075406-10G 075406-100G
102541-208EA 72.31 USD
102541-208 102541-210 102541-212
Thymidine (2'-deoxythymidine) ≥95%
Thymidine (2'-Deoxythymidine)

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₅
MDL Number: MFCD00006537
CAS Number: 50-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR