Uridine ≥95%

Supplier: Matrix Scientific

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside

075397-100G 075397-25G
102541-168EA 75.38 USD
102541-168 102664-042
Uridine ≥95%
Uridine

Hazard: Irritant

Formula: C₉H₁₂N₂O₆
MW: 244.20 g/mol
Melting Pt: 163…167 °C
MDL Number: MFCD00006526
CAS Number: 58-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR