Methyl-5-hydroxynicotinate ≥95%

Supplier: Matrix Scientific

075356-5G 075356-25G
102540-980EA 155.38 USD
102540-980 102551-958
Methyl-5-hydroxynicotinate ≥95%
Methyl 5-hydroxynicotinate

Hazard: Irritant

Formula: C₇H₇NO₃
Melting Pt: 195…197 °C
MDL Number: MFCD00191521
CAS Number: 30766-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR