(S)-(+)-2-Aminobutyric acid ≥95%

Supplier: Matrix Scientific

Synonyms: L-2-Aminobutyric acidL-alpha-Amino-N-Butyric acid(S)-2-Aminobutanoic acidH-Abu-OH

075329-100G
102540-864EA 89.23 USD
102540-864
(S)-(+)-2-Aminobutyric acid ≥95%
(S)-(+)-2-Aminobutyric acid

Hazard: Irritant

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00064415
CAS Number: 1492-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR