Ethanesulfonamide ≥95%

Supplier: Matrix Scientific

075319-1G 075319-5G
102540-824EA 61.54 USD
102540-824 102540-826
Ethanesulfonamide ≥95%
Ethanesulfonamide

Hazard: Irritant

Formula: C₂H₇NO₂S
MW: 109.1475 g/mol
Melting Pt: 57…60 °C
MDL Number: MFCD03265410
CAS Number: 1520-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR