Benzocaine (ethyl-4-aminobenzoate) ≥95%

Supplier: Matrix Scientific

Synonyms: AethoformEthyl 4-aminobenzoate

075318-100G
102540-822EA 29.23 USD
102540-822
Benzocaine (ethyl-4-aminobenzoate) ≥95%
Benzocaine (Ethyl 4-aminobenzoate)

Hazard: Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 88…92 °C
MDL Number: MFCD00007892
CAS Number: 94-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR