(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride
, H-Lys(Z)-OBzl·HCl
, H-Lys(Z)-OBzl.HCl
075233-1G
075233-5G
075233-10G
102540-428EA
284.62
USD
102540-428
102540-432
102540-430
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride ≥95%
(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Hazard: Irritant
Formula:
C₂₁H₂₆N₂O₄·HCl MW: 406.9 g/mol |
MDL Number:
MFCD00038981 CAS Number: 6366-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...