L-Lysine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH

075211-100G 075211-500G
102540-318EA 116.92 USD
102540-318 102551-944
L-Lysine ≥95%
L-Lysine

Hazard: Irritant

Formula: C₆H₁₄N₂O₂
MW: 146.19 g/mol
Melting Pt: 215 °C
MDL Number: MFCD00064433
CAS Number: 56-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR