2-Amino-5-chlorobenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloroanthranilonitrile

075060-5G 075060-10G 075060-100G
102539-554EA 72.31 USD
102539-554 102539-556 102539-558
2-Amino-5-chlorobenzonitrile ≥95%
2-Amino-5-chlorobenzonitrile

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.58
Boiling Pt: 101
MDL Number: MFCD00017106
CAS Number: 5922-60-1
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR