L-Valine-tert-butyl ester hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
H-Val-OtBu·HCl
, L-2-Aminoisovaleric Acid tert-Butyl Ester Hydrochloride
, (S)-tert-Butyl 2-amino-3-methylbutanoate hydrochloride
074855-10G
074855-100G
102538-560EA
387.69
USD
102538-560
102538-558
L-Valine-tert-butyl ester hydrochloride ≥95%
L-Valine-tert-butyl ester hydrochloride
Hazard: Irritant
Formula:
C₉H₁₉NO₂·HCl MW: 209.7 g/mol |
MDL Number:
MFCD00034863 CAS Number: 13518-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...