8-Bromo-3-iodoimidazo[1,2-a]pyrazine ≥95%
Supplier: Matrix Scientific
074641-250MG
074641-1G
102537-470EA
826.15
USD
102537-470
102537-472
8-Bromo-3-iodoimidazo[1,2-a]pyrazine ≥95%
8-Bromo-3-iodoimidazo[1,2-a]pyrazine
Hazard: Irritant
Formula:
C₆H₃BrIN₃ |
CAS Number:
1245644-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...