(S)-1-Boc-3-hydroxypiperidine ≥95%
Supplier: Matrix Scientific
Synonyms:
(S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, (3S)-3-Hydroxypiperidine, N-BOC protected
074558-5G
074558-1G
074558-25G
102537-046EA
16.92
USD
102537-046
102664-010
102664-012
(S)-1-Boc-3-hydroxypiperidine ≥95%
(S)-1-Boc-3-hydroxypiperidine
Hazard: Irritant
Formula:
C₁₀H₁₉NO₃ |
MDL Number:
MFCD04115307 CAS Number: 143900-44-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...