3-Nitro-p-toluic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-3-nitrobenzoic acid

074487-100G
102536-676EA 155.38 USD
102536-676
3-Nitro-p-toluic acid ≥95%
3-Nitro-p-toluic acid

Hazard: Irritant

Formula: C₈H₇NO₄
MW: 181.15 g/mol
MDL Number: MFCD00007174
CAS Number: 96-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR